Learn More
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Specifications
Specifications
| Melting Point | 500.0°C |
| Color | Orange |
| Physical Form | Powder |
| Quantity | 100 g |
| Assay Percent Range | 97.5% min. (PB3O4) |
| Linear Formula | 2PbO·PbO2 |
| Merck Index | 15,548 |
| Solubility Information | Solubility in water: insoluble. Other solubilities: partly soluble in acids, insoluble in alcohol |
| Formula Weight | 685.57 |
| Percent Purity | 98% |
| Show More |
Safety and Handling
- Lead(II,IV) oxide
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 2
- Reproductive toxicity on or via lactation
- Reproductive toxicity Category 1B
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H351-Suspected of causing cancer.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H362-May cause harm to breast-fed children.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P263-Avoid contact during pregnancy/while nursing.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contain: Lead oxide
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.