missing translation for 'onlineSavingsMsg'
Learn More

Lead(II,IV) oxide, red, 98%

Codice prodotto. 10430881 Sfoglia Tutto Thermo Scientific Chemicals Prodotti
Cambia vista
Click to view available options
Quantity:
5 g
100 g
500 g
5 kg
Packaging:
Glass bottle
Dimensione della confezione:
100g
500g
5g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity Packaging unitSize
10430881 500 g Glass bottle 500g
10781301 5 g Glass bottle 5g
10626052 100 g Glass bottle 100g
15159243 5 kg Glass bottle 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 10430881 Fornitore Thermo Scientific Acros N. del fornitore 221115000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 1314-41-6 | O4Pb3 | 685.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 1314-41-6
Molecular Formula O4Pb3
Molecular Weight (g/mol) 685.57
MDL Number MFCD00078491
InChI Key XMFOQHDPRMAJNU-UHFFFAOYSA-N
Synonym lead tetroxide, orange lead, lead oxide, red, lead ii,iv oxide, po4b3, lead oxide, red, lr, lead oxide, red powder, 1-2 mum, lead oxide, red, saj first grade, lead oxide, powder trace metals basis
PubChem CID 16685188
IUPAC Name 1,3,5,7-tetraoxa-2$l^{2},4,6$l^{2}-triplumbaspiro[3.3]heptane
SMILES O1[Pb]O[Pb]12O[Pb]O2

Specifica

Melting Point 500.0°C
Color Orange
Physical Form Powder
Quantity 500 g
Assay Percent Range 97.5% min. (PB3O4)
Linear Formula 2PbO·PbO2
Merck Index 15,548
Solubility Information Solubility in water: insoluble. Other solubilities: partly soluble in acids, insoluble in alcohol
Formula Weight 685.57
Percent Purity 98%
Packaging Glass bottle
Chemical Name or Material Lead(II, IV) oxide, red, 98%
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.