missing translation for 'onlineSavingsMsg'
Learn More

Lead(II,IV) oxide, 97% (metals basis), Thermo Scientific Chemicals

Catalog Number 11377797
Click to view available options
Quantity:
1 kg
5 kg
This item is not returnable. View return policy
1314-41-6
O4Pb3
685.596
MFCD00078491
XMFOQHDPRMAJNU-UHFFFAOYSA-N
lead tetroxide, orange lead, lead oxide, red, lead ii,iv oxide, po4b3, lead oxide, red, lr, lead oxide, red powder, 1-2 mum, lead oxide, red, saj first grade, lead oxide, powder trace metals basis
16685188
1,3,5,7-tetraoxa-2$l^{2},4,6$l^{2}-triplumbaspiro[3.3]heptane
O1[Pb]O[Pb]12O[Pb]O2
This item is not returnable. View return policy
1314-41-6
O4Pb3
685.596
MFCD00078491
XMFOQHDPRMAJNU-UHFFFAOYSA-N
lead tetroxide, orange lead, lead oxide, red, lead ii,iv oxide, po4b3, lead oxide, red, lr, lead oxide, red powder, 1-2 mum, lead oxide, red, saj first grade, lead oxide, powder trace metals basis
16685188
1,3,5,7-tetraoxa-2$l^{2},4,6$l^{2}-triplumbaspiro[3.3]heptane
O1[Pb]O[Pb]12O[Pb]O2

Lead(II,IV) oxide is used to prepare colorless glass, faience glaze, porcelain painting flux, iron and steel coatings, rubber pigment and in glass cement. It is also used in gas and steam pipes, storage batteries, writing on glass and to make lead peroxide and matches. It is associated with linseed oil and used as a thick, long-lasting anti-corrosive paint. It gives better water resistant properties by replacing magnesium oxide. Further, it is used in the manufacture of lead glass and rustproof primer paints. In addition to this, it acts as a pigment for primer paints for iron objects.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point ∼500°C (decomposition)
Color Orange to Red
Physical Form Powder
Quantity 1 kg
Linear Formula Pb3O4
UN Number UN1479
Merck Index 14,5425
Solubility Information Soluble in hydrochloric acid,glacial acetic acid and nitric acid and hydrogen peroxide. Insoluble in water and alcohol.
Formula Weight 685.6
Percent Purity 97%
Odor Odorless
Density 9.1 g/mL
Assay (metals basis)
Chemical Name or Material Lead(II,IV) oxide
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead(II,IV) oxide
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 2
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'