missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) fluoride, 99% min

Product Code. 11369229
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11369229 250 g 250g
11379229 50 g 50g
11389229 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11369229 Supplier Thermo Scientific Alfa Aesar Supplier No. 088282.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7783-46-2 | F2Pb | 245.20 g/mol

Lead(II) fluoride is a specialty product for proteomics research. It is used in low melting glasses, in glass coatings to reflect infrared rays, in phosphors for television-tube screens and as a catalyst for the manufacture of picoline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-46-2
Molecular Formula F2Pb
Molecular Weight (g/mol) 245.20
MDL Number MFCD00011162
InChI Key FPHIOHCCQGUGKU-UHFFFAOYSA-L
Synonym lead fluoride, lead ii fluoride, lead difluoride, lead fluoride pbf2, plumbous fluoride, lead +2 fluoride, difluoroplumbane, plomb fluorure french, pbf2, plomb fluorure
PubChem CID 24549
IUPAC Name difluorolead
SMILES [F-].[F-].[Pb++]

Specifications

Boiling Point 1290°C
Melting Point 855°C
Physical Form Powder
Quantity 250 g
UN Number UN2291
Merck Index 14,5408
Solubility Information Sparingly soluble in water(0.64g/L at 20°C). Soluble in nitric acid. Insoluble in acetone and ammonia.
Formula Weight 245.2
Percent Purity ≥99%
Odor Odorless
Density 8.445 g/mL
Chemical Name or Material Lead (II) fluoride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead(II) fluoride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Lead fluoride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.