missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) chloride, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number 11340058
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
7758-95-4
Cl2Pb
278.10
MFCD00011157
HWSZZLVAJGOAAY-UHFFFAOYSA-L
lead dichloride, lead ii chloride, lead chloride, plumbous chloride, lead 2+ chloride, lead chloride pbcl2, pbcl2, ccris 7565, lead ii chloride, ultra dry
24459
[Cl-].[Cl-].[Pb++]
This item is not returnable. View return policy
7758-95-4
Cl2Pb
278.10
MFCD00011157
HWSZZLVAJGOAAY-UHFFFAOYSA-L
lead dichloride, lead ii chloride, lead chloride, plumbous chloride, lead 2+ chloride, lead chloride pbcl2, pbcl2, ccris 7565, lead ii chloride, ultra dry
24459
[Cl-].[Cl-].[Pb++]

Used in the synthesis of lead titanate and barium lead titanate ceramics; employed in the production of infrared transmitting glass and ornamental glass (aurene glass); useful as an electrode in geophysical applications and in solar cells.

Lead chloride is used in the synthesis of lead titanate and barium lead titanate ceramics; employed in the production of infrared transmitting glass and ornamental glass (aurene glass); useful as an electrode in geophysical applications and in solar cells. Analytical reagent, preparation of lead salts, as solder and flux. Pattinson's white lead, pigment in white paint, is the basic lead chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 951°C
Melting Point 501°C
Color White
Physical Form Powder
Quantity 2 g
Assay Percent Range 99.999% (Metal basis)
Solubility Information Soluble in hot water,alkali hydroxides and ammonium chloride solution. Insoluble in cold water and alcohol.
IUPAC Name λ²-lead(2+) dichloride
Formula Weight 278.1
Odor Odorless
Appearance White powder
Density 5.85 g/cm3 at 20°C
Chemical Name or Material Lead(II) chloride, ultra dry
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead(II) chloride, ultra dry
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P201-Obtain special instructions before use.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'