Learn More
Lead(II) carbonate, basic, 99% (metals basis), Thermo Scientific Chemicals
Chemical Identifiers
CAS | 1319-46-6 |
---|---|
Molecular Formula | C2H2O8Pb3 |
MDL Number | MFCD00078155 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11389727
|
Thermo Scientific Alfa Aesar
042089.22 |
100 g |
N/A
|
N/A | |||||
11309737
|
Thermo Scientific Alfa Aesar
042089.36 |
500 g |
N/A
|
N/A | |||||
11399727
|
Thermo Scientific Alfa Aesar
042089.A3 |
2 kg |
N/A
|
N/A | |||||
Description
Lead(II) carbonate is employed in the production of anticorrosion paint and outdoor paint. It is also used as a precursor for cosmetics, colored glaze and dope. It finds application in the tin-lead plating. It is also useful as a stabilizing agent for vinyl chloride plastic.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1319-46-6 | |
C2H2O8Pb3 | |
MFCD00078155 |
Specifications
1319-46-6 | |
Powder | |
C2H2O8Pb3 | |
White lead, C.I. 77597 | |
White powder | |
Lead(II) carbonate, Basic |
White | |
99% (Metal basis) | |
MFCD00078155 | |
775.63 | |
6.14g/cm3 at 20°C |
Safety and Handling
H301-H332-H360-H373-H400-H410: Toxic if swallowed. Harmful if inhaled. May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
missing translation for 'dotInformation' : DOT Class: 9, Packaging Group: III
missing translation for 'einecsNumber' : 215-290-6
- Lead(II) carbonate, basic
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P201-Obtain special instructions before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only