missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead(II) carbonate, ACS, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
4 x 500 g
Description
Lead(II) carbonate is used in polymerization catalysts. It serves as a curing agent for silicon resins, heat-resistant lubricants, lead-acid storage batteries, wire-reinforced hoses, thermistors and heat stabilizers for polyvinyl chloride polymers. Further, it is used in electrophotography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White |
Physical Form | Powder |
Quantity | 4 x 500 g |
Solubility Information | Soluble in acid and alkali. Insoluble in water,alcohol and ammonia. |
IUPAC Name | λ²-lead(2+) carbonate |
Formula Weight | 267.2 |
Grade | ACS |
Odor | Odorless |
Appearance | White powder |
Chemical Name or Material | Lead(II) carbonate, For analysis ACS |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'