missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) Carbonate, ACS Reagent, Honeywell™ Fluka™

Product Code. 15654020 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15654020 100 g Poly Bottle 100g
15664020 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15654020 Supplier Honeywell-Fluka Supplier No. 336378100G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 598-63-0
Molecular Formula CO3Pb
Molecular Weight (g/mol) 267.20
MDL Number MFCD00064631
InChI Key MFEVGQHCNVXMER-UHFFFAOYSA-L
Synonym lead carbonate, lead ii carbonate, cerussete, cerussite, lead 2+ carbonate, plumbous carbonate, dibasic lead carbonate, unii-43m0p24l2b, carbonic acid, lead 2+ salt 1:1, lead carbonate pbco3
PubChem CID 11727
SMILES [Pb++].[O-]C([O-])=O

Specifications

Melting Point 400°C
Color White
Quantity 100 g
Linear Formula PbCO3
UN Number UN3077
IUPAC Name λ²-lead(2+) carbonate
Formula Weight 267.21g/mol
Packaging Poly Bottle
Appearance Powder
Density 6.140g/cm3
Chemical Name or Material Lead(II) carbonate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.