Learn More
Lead(II) bromide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals
Use promo code "21615" to get your promotional price.
Description
Lead(II) bromide is used in the field of antirust, pigment and photograph. The molten lead(II) bromide acts as an electrolyte. Lead(II) bromide provides a high concentration of lead(II) ions and bromide ions to carry the current during the electrolysis process. The rare-earth-doped alkali-lead bromide crystals (potassium lead bromide (or) KPB; rubidium lead bromide or RPB) emerge as promising new low-phonon-energy host materials for mid-IR applications and are useful for solid state lasers. Hybrid organic/lead halide perovskites are promising materials for solar cell fabrication.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 916°C |
Melting Point | 373°C |
Color | White |
Physical Form | Powder |
Quantity | 25 g |
Assay Percent Range | 99.999% (Metals basis) |
Solubility Information | Slightly soluble in water. Insoluble in alcohol. Soluble in ammonia and alkali. |
IUPAC Name | λ²-lead(2+) dibromide |
Formula Weight | 367.01 |
Odor | Odorless |
Show More |
RUO – Research Use Only