missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) bromide, Puratronic™, 99.998% (metals basis)

Product Code. 11396787
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11396787 25 g 25g
11386787 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11396787 Supplier Thermo Scientific Alfa Aesar Supplier No. 010720.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lead(II) bromide, 99.999%, Br2Pb, CAS Number-10031-22-8, dibromoplumbane, acmc-1bq2x, lead 2+ bromide, ccris 4220, lead dibromide, lead bromide pbbr2, lead ii bromide, pbbr2, leadbromide, lead bromide, 25g, (metals basis), 916 deg.C, White, 6.66 g/mL | CAS: 10031-22-8 | Br2Pb | 367.00 g/mol

It is used in the field of antirust, pigment and photograph. The molten lead(II) bromide acts as an electrolyte. Lead(II) bromide provides a high concentration of lead(II) ions and bromide ions to carry the current during the electrolysis process. The rare-earth-doped alkali-lead bromide crystals (potassium lead bromide (or) KPB; rubidium lead bromide or RPB) emerge as promising new low-phonon-energy host materials for mid-IR applications and are useful for solid state lasers. Hybrid organic/lead halide perovskites are promising materials for solar cell fabrication.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10031-22-8
Molecular Formula Br2Pb
Molecular Weight (g/mol) 367.00
MDL Number MFCD00011156
InChI Key ZASWJUOMEGBQCQ-UHFFFAOYSA-L
Synonym lead ii bromide, lead bromide, lead dibromide, lead bromide pbbr2, pbbr2, lead 2+ bromide, dibromoplumbane, ccris 4220, leadbromide, acmc-1bq2x
PubChem CID 24831
IUPAC Name dibromolead
SMILES [Br-].[Br-].[Pb++]

Specifications

Boiling Point 916°C
Melting Point 373°C
Color White
Physical Form Powder
Quantity 25 g
Linear Formula PbBr2
UN Number UN2291
Merck Index 14,5403
Solubility Information Slightly soluble in water; Insoluble in alcohol; Soluble in ammonia,alkali,KBr and NaBr.
Formula Weight 367.01
Percent Purity 99.999%
Grade Puratronic
Odor Odorless
Density 6.66 g/mL
Assay (metals basis)
Chemical Name or Material Lead(II) bromide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead(II) bromide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contain: lead bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.