missing translation for 'onlineSavingsMsg'
Learn More

Lead(Ii) Acetate Trihydrate, puriss. p.a. ACS Reagent, Solstice

Product Code. 15643800 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
Unit Size:
100g
1kg
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15643800 250 g 250g
15623800 100 g 100g
15633800 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15643800 Supplier Honeywell-Fluka Supplier No. 32307250G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

Puriss. p.a., ACS Reagent, Reagent ISO, Reagent European Pharmacopoeia, 99.5-102.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 6080-56-4
Molecular Formula C4H12O7Pb
Molecular Weight (g/mol) 379.30
MDL Number MFCD00150023
InChI Key MCEUZMYFCCOOQO-UHFFFAOYSA-L
Synonym lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate
PubChem CID 22456
ChEBI CHEBI:33112
IUPAC Name lead(2+);diacetate;trihydrate
SMILES O.O.O.[Pb++].CC([O-])=O.CC([O-])=O

Especificaciones

Quantity 250 g
Linear Formula Pb(CH3CO2)2 · 3H2O
UN Number 1616
Beilstein 3730298
Formula Weight 379.33g/mol
Percent Purity 99.5 to 102%
Grade ACS/puriss. p.a.
Chemical Name or Material Lead(Ii) Acetate Trihydrate, Reag. Ph. Eur. reag. ISO
compliance-icons compliance-icons
Nombre del producto
  • Lead(II) acetate trihydrate
Palabra de advertencia
  • Danger
Clasificación
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
  • Acute toxicity Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Indicaciones de peligro
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Consejos de prudencia
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.