Learn More
Lead(II) acetate trihydrate, Puratronic™, 99.995% (metals basis)
Description
Lead(II) acetate trihydrate is used as a mordant in textile printing and dyeing, drier in paints, varnishes and as a water repellant. It is also used in cosmetics and to prepare other lead compounds. It serves as an ingredient in progressive types of hair coloring dyes. Further, it is used in the detection of poisonous gas hydrogen sulfide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 75°C |
| Physical Form | Crystalline |
| Quantity | 250 g |
| UN Number | UN1616 |
| Beilstein | 3730298 |
| Merck Index | 14,5397 |
| Solubility Information | Freely soluble in water,alcohol and glycerol. |
| Formula Weight | 379.33 (325.28 Anhydrous) |
| Percent Purity | 99.995% |
| Grade | Puratronic |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.