missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead(II) acetate trihydrate, 99+%, for analysis, Thermo Scientific Chemicals
13.40 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 75.0°C |
Color | White |
Physical Form | Crystalline Powder and/or Crystals |
Quantity | 25 g |
Assay Percent Range | 99.0 to 103% (Complexometry) |
Linear Formula | (CH3CO2)2Pb·3H2O |
Merck Index | 15, 5452 |
Solubility Information | Solubility in water: 625g/L. Other solubilities: 1g/30mL alcohol, freely soluble in glycerol |
IUPAC Name | λ²-lead(2+) diacetate trihydrate |
Formula Weight | 379.33 |
Show More |
Safety and Handling
Signal Word
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- H318-Causes serious eye damage.
- H350-May cause cancer.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'