missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) acetate basic, ACS reagent

Product Code. 15254288 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15254288 1 kg 1kg
15264288 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15254288 Supplier Thermo Scientific Acros Supplier No. 453220010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 51404-69-4 | C4H10O8Pb3 | 807.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51404-69-4
Molecular Formula C4H10O8Pb3
Molecular Weight (g/mol) 807.69
InChI Key XAOPZCQFBUCRKR-UHFFFAOYSA-L
PubChem CID 75087083
IUPAC Name diacetyloxylead;lead;dihydrate
SMILES CC(=O)O[Pb]OC(=O)C.O.O.[Pb]

Specifications

Quantity 1 kg
Formula Weight 807.69
Percent Purity ≥33.0% (basic Pb as PbO)
Grade ACS Reagent
Loss on Drying 1.5% max. (105°C, 2 h)
Chemical Name or Material Lead(II) acetate basic
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead(II) acetate basic
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.