missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) acetate, basic, ACS

Product Code. 11327858
Change view
Click to view available options
Quantity:
500 g
2 kg
Unit Size:
2kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11327858 2 kg 2kg
11337858 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11327858 Supplier Thermo Scientific Alfa Aesar Supplier No. 036393.A3
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1335-32-6

Lead(II) acetate is used in the production of sweeteners, cosmetics, astringent, used in the cleaning and maintenance of stainless steel firearm suppressors (silencers) and compensators, and also used in making of slow matches.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1335-32-6
MDL Number MFCD00013041
Synonym Lead subacetate

Specifications

Physical Form Powder
Quantity 2 kg
Linear Formula Pb(OOCCH3)2·2Pb(OH)2
UN Number UN1616
Merck Index 14,5419
Solubility Information Soluble in water.
Formula Weight 807.71
Grade ACS
Sensitivity Air Sensitive
Chemical Name or Material Lead(II) acetate, Basic

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.