missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Lead granules, approximately 3mm (0.12 in.), 99.9% (metals basis)

Product Code. 11324647
Change view
Click to view available options
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11324647 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11324647 Supplier Alfa Aesar™ Supplier No. 039685.18

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Color Gray
CAS 7439-92-1
Quantity 50 g
Odor Odorless
Form Granular
Molecular Formula Pb
MDL Number MFCD00134050
Synonym plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant
InChI Key WABPQHHGFIMREM-UHFFFAOYSA-N
SMILES [Pb]
IUPAC Name lead
Molecular Weight (g/mol) 207.20
PubChem CID 5352425
ChEBI CHEBI:27889
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Lead granules, Approximately 3mm (0.12 in.)
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead granules
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to accordance with local/regional/national/international regulations..
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.