Learn More
Thermo Scientific Alfa Aesar Lead granules, approximately 3mm (0.12 in.), 99.9% (metals basis)
Brand: Thermo Scientific Alfa Aesar 039685.30
Lead granules, approximately 3mm (0.12 in.), 99.9% (metals basis)
Code : NEW
Additional Details : CAS Number : 7439-92-1 Weight : 0.25000kg