missing translation for 'onlineSavingsMsg'
Learn More

Lead-Free Paint (Powdered) Reference Material: Pb < 0.01 μg/g

Product Code. 30309970
Change view
Click to view available options
Quantity:
20 g
Unit Size:
20g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30309970 20 g 20g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30309970 Supplier VHG Labs Supplier No. VHGPBFP20G
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cadmium∣Chromium∣Lead
CAS 7440-43-9, 7440-47-3, 7439-92-1
Analyte or Component Names Cadmium∣Chromium∣Lead
Shipping Condition Room Temperature
Solution Type Matrix Material
Element Multi-element
Solvent or Matrix Paint
Industry Type Petrochemical Testing
Quantity 20 g
Concentration or Composition (by Analyte or Components) Lead <0.01 μg/g∣Cadmium <0.1 mg/kg∣Chromium <0.1 mg/kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.