Learn More
STERIS™ Lead-free Autoclave Indicator Tape
Seal products and packages for sterilization with this self-adhering tape. STERIS™ Lead-free Autoclave Indicator Tape features green lead-free stripes that change color to brown/black providing an immediate indication that the sterilization process is complete.
£17.65 - £23.35
Product Code | Brand | Dimensions (LxW) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (LxW) | Price | Quantity & Availability | |||||
12370489
|
STERIS™
0162AB |
50 m x 19 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12739135
|
STERIS™
0163AB |
50 m x 25 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Self-adhering tape with a chemical indicator printed on one side
- The easily recognizable stripes change color when exposed to the specific sterilization, verifying exposure to the process without the need to open packs or check records
- Color change brown/black
Specifications
Brown, Black |