missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead foil, 0.5mm (0.02in) thick, Puratronic, 99.99% (metalsbasis)
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Gray |
| Quantity | 50 x 50 mm |
| Odor | Odorless |
| Weight | ≈3.5g/25x25mm |
| Form | Foil |
| Percent Purity | 99.998% |
| Grade | Puratronic |
| Assay | (metals basis) |
| Chemical Name or Material | Lead foil, 0.5mm (0.02 in.) thick |
Safety and Handling
Signal Word
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 1
- Reproductive toxicity on or via lactation
- Reproductive toxicity Category 1A
- Specific target organ toxicity Category 1
- H360FD-May damage fertility. May damage the unborn child.
- H362-May cause harm to breast-fed children.
- H372-Causes damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?