Learn More
Lead Antimony shot, 2mm (0.08in), Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 010224.36
Code : NEW
Additional Details : CAS Number : 7439-92-1 Weight : 0.50000kg Transport : UN number : 3077 Chem class : 9 Pack group : III
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Lead Antimony shot | |
MFCD00134050 | |
500 g | |
Shot |
2mm (0.08 in.) | |
UN3077 | |
Odorless |
Safety and Handling
- Lead Antimony shot
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only