missing translation for 'onlineSavingsMsg'
Learn More

Lead antimonide, 99+% (metals basis), Thermo Scientific™

Product Code. 11383438
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11383438 5 g 5g
11373438 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11383438 Supplier Thermo Scientific Alfa Aesar Supplier No. 036350.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12266-38-5
Molecular Formula PbSb
Molecular Weight (g/mol) 328.96
MDL Number MFCD00151190
InChI Key QQHJESKHUUVSIC-UHFFFAOYSA-N
Synonym lead monoantimonide, lead antimonide, lead-antimonide, stibanylidyneplumbyl, antimony, compd. with lead 1:1, antimony, compound with lead 1:1
PubChem CID 72720464
IUPAC Name stibanylidynelead
SMILES [Sb]#[Pb]

Specifications

Physical Form Lump
Quantity 5 g
UN Number UN1549
Formula Weight 328.94
Percent Purity ≥99%
Odor Odorless
Density 6.72 g/mL
Assay (metals basis)
Chemical Name or Material Lead antimonide
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead antimonide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.