missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead, Alloy, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Lead, Alloy, Ø40x15mm
Specifications
Specifications
| Chemical Name or Material | Sulfur/Iron/Zinc/Cadmium/Nickel/Tellurium/Silver/Bismuth/Selenium/Copper/Tin/Arsenic/Antimony/Lead |
| CAS | 7439-92-1 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Sulfur -20 ppm/Iron 0.0002 %/Zinc 0.0002 %/Cadmium 0.00027 %/Nickel 0.00032 %/Tellurium 0.0006 %/Silver 0.0011 %/Bismuth 0.0118 %/Selenium 0.016 %/Copper 0.0178 %/Tin 0.024 %/Arsenic 0.057 %/Antimony 2.89 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction