missing translation for 'onlineSavingsMsg'
Learn More

Lead Acetate Trihydrate, TRC

Product Code. 30475298
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30475298 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30475298 Supplier Toronto Research Chemicals Supplier No. TRCL32006010G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lead Acetate Trihydrate
CAS 6080-56-4
Molecular Formula C4H6O4Pb . 3H2O
Quantity 10 g
Synonym Lead(II) Acetate Trihydrate
InChI Formula InChI=1S/2C2H4O2.3H2O.Pb/c2*1-2(3)4;;;;/h2*1H3,(H,3,4);3*1H2;/q;;;;;+2/p-2
SMILES O.O.O.[Pb+2].CC(=O)[O-].CC(=O)[O-]
IUPAC Name lead(2+);diacetate;trihydrate
Molecular Weight (g/mol) 379.33
Formula Weight 380.0349
Product Title
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.