missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| CAS | 301-04-2 |
| Molecular Formula | C4H6O4Pb |
| Quantity | 50 mL |
| Synonym | lead acetate, lead diacetate, lead ii acetate, lead 2+ acetate, unichem pba, lead di acetate, normal lead acetate, dibasic lead acetate, lead dibasic acetate, bleiacetat german |
| InChI Key | GUWSLQUAAYEZAF-UHFFFAOYSA-L |
| Molecular Weight (g/mol) | 325.30 |
| PubChem CID | 18668094 |
Safety and Handling
Product Identifier
- Lead Acetate Paper. European Pharmacopoeia Ref: 1048102 (RP 112)
- Danger
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- LONG-TERM AQUATIC HAZARD Chronic 2
- H360-May damage fertility or the unborn child.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P314-Get medical advice/attention if you feel unwell.
- P391-Collect spillage.
- P405-Store locked up.
- P501-Dispose of contents/container to .
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
