missing translation for 'onlineSavingsMsg'
Learn More

Emma Safety Footwear Le Mans S1P Shoe

Product Code. 17592476
Change view
Click to view available options
Size:
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17592476 39 1 pair
17502486 40 1 pair
17512486 41 1 pair
17522486 42 1 pair
17532486 43 1 pair
17542486 44 1 pair
17552486 45 1 pair
17562486 46 1 pair
17572486 47 1 pair
17582486 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Product Code. 17592476 Supplier Emma Safety Footwear Supplier No. MM70354739
  • ONLINE EXCLUSIVE PRICE
    £126.00
    £94.76 / 1 pairs
    Save £31.24
    24% Off
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Like its name suggests, you could wear the Le Mans for 24 hr. as the shoes offer optimal ventilation, perspiration and moisture removal with the Hydro-Tec™ antibacterial lining material.

  • Metal Free
  • Dry, fresh feet for up to 24 hr.The name already suggests it: you can safely wear LeMans for 24 hours.
  • The shoe offers optimal ventilation and moisture is immediately removed by the Hydro-Tec™ antibacterial lining.
  • LeMans is ideal for indoor working environments and work in transport and logistics.
  • LeMans looks sporty, that's how the shoe feels; soles with energy return and a fit that hugs the feet perfectly.
  • Industries:
    • Logistics & Transport
    • Electronics
    • Light Industry
  • Safety shoe
    • S1P-Safety shoe for dry conditions

Specifications

Certifications/Compliance ISO 20345:2011
Closure Type Laces
Color Black, Gray, Orange
For Use With (Application) Logistics and Transport, Electronics, Manufacturing
Liner Material Hydro-Tec Antibacterial
Size 39
Fit Standard Width
Ankle Style Low-cut
Product Type Safety Shoe
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.