missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals LDS-sample buffer (4X), non-reducing

Product Code. 15474379
Change view
Click to view available options
Quantity:
25 mL
50 mL
Unit Size:
25mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15474379 25 mL 25mL
15484379 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15474379 Supplier Thermo Scientific Chemicals Supplier No. J61894.AC
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LDS-sample buffer (4X), non-reducing is used in SDS-polyacrylamide gel electrophoresis (SDS-PAGE). It is used in denaturing gels and is compatible with both coomassie and silver staining and Western blotting applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
LDS-sample buffer (4X), non-reducing is used in SDS-polyacrylamide gel electrophoresis (SDS-PAGE). It is used in denaturing gels and is compatible with both coomassie and silver staining and Western blotting applications.

Solubility
Not miscible in water.

Notes
This buffer contains: 988mM Tris-HCl, 2.04mM EDTA, 8% LDS, 40% glycerol, 0.88% Coomassie blue and 0.7mM phenol red at pH 8.5.
TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Concentration 4X
Physical Form Liquid
Quantity 25 mL
At what temperature should I store LDS-sample buffer (4X), non-reducing (Cat. No. J61894.xx)?

LDS-sample buffer (4X), non-reducing should be stored at room temperature (20-25 degrees Celsius) upon receipt.

What protocol should I use for LDS-sample buffer (4X), non-reducing (Cat. No. J61894.xx)?

We recommend the following protocol for use of LDS-sample buffer (4X), non-reducing:
1. Mix one part LDS sample buffer to three parts protein (e.g., 5 µL sample buffer + 15 µL protein sample). Failure to properly dilute may result in a diffuse dye front.
2. Heat mixture at 100°C for 3-5 minutes.
3. Allow sample to cool back to room temperature.
4. Load sample onto SDS-polyacrylamide gel and begin electrophoresis.
5. Stop electrophoresis when the dye front nears the bottom of the gel.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.