missing translation for 'onlineSavingsMsg'
Learn More

Melford LDS

Product Code. 30018431
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018431 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30018431 Supplier Melford Supplier No. L2620025.0
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lithium Dodecyl Sulfate (LDS) is a chemical compound with significant applications in the life sciences, particularly in molecular biology and protein research.

It is a type of anionic detergent and belongs to the class of alkyl sulfates.

LDS is commonly used as a surfactant and denaturing agent in various laboratory techniques. It is widely used in protein electrophoresis, particularly in sodium dodecyl sulfate-polyacrylamide gel electrophoresis (SDS-PAGE).

LDS helps solubilize proteins, denature them, and impart a negative charge proportional to their size, allowing for separation based on molecular weight. It is also used in membrane protein studies and as a component in protein extraction buffers. It aids in the solubilization and extraction of membrane proteins, facilitating their analysis and characterization.

Additionally, LDS is utilized in the preparation of protein samples for mass spectrometry analysis. It helps to enhance protein solubility, reduce protein-protein interactions, and promote efficient digestion of proteins by proteases.

Specifications

CAS 2044-56-6
Molecular Formula C12H25LiO4S
Quantity 25 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.