missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ LDPE Wash Bottles with Venting Valve
Shop All Brand Products
Click to view available options
Capacity (Metric):
250 mL
500 mL
Chemical Label:
Acetone
Distilled Water
Ethanol
Isopropanol
Methanol
Unit Size:
Pack of 5
Description
- For distilled water and organic solvents only
- Large opening for convenient filling
- Label with designation of contents and general safety statements in English (was CAS numbers and NFPA codes) and German (was CAS numbers)
- Manufactured from flexible PE-LD, limited to temperatures below 80°C
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| Material | LDPE |
| Closure Color | Blue |
| Closure Included | Yes |
| Mouth | Wide |
| Product Type | Wash Bottle |
| Chemical Label | Isopropanol |
| Closure Type | Stopper |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction