missing translation for 'onlineSavingsMsg'
Learn More

Kautex™ 500 mL Safety Wash Bottles Series 303 LDPE

Product Code. 15768877
Click to view available options
Chemical Label:
Acetone
Distilled Water
Ethanol
Isopropanol
Methanol
Closure Color:
Blue
Green
Orange
Red
Yellow
Unit Size:
Each
Pack of 10
Pack of 100

Product Code. 15768877

Brand: Kautex™ 2000770021

Only null left
Add to basket
Add to basket

Designed to give an accurate fine controlled stream of liquid. Kautex™ 500mL Safety Wash Bottles provide excellent chemical resistance for laboratory safety.

  • Imprinted with hazard symbols and substances of content in German, English and French language
  • Writing space
  • Abrasion-resistant printing
  • Closure color matches color of ink for easy identification of media
  • Leak-proof
  • Completely emptied by sliding delivery tube, even when inverted.
Suitable for the declared liquids. Not suitable for food.
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 500 mL
Color Natural
Diameter (Metric) Inner Neck 39.2 mm
Material LDPE
No. per Pack 10
Quantity 100 Pk.
Closure Color Green
Mouth Wide
Shape Round
Height (Metric) 154 mm
Product Type Wash Bottle
Chemical Label Ethanol
Diameter (Metric) 77 mm
Closure Type Screw Cap with Delivery Nozzle
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.