missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kautex™ 250 mL Safety Wash Bottles Series 303 LDPE
Click to view available options
Chemical Label:
Acetone
Distilled Water
Ethanol
Isopropanol
Methanol
Closure Color:
Blue
Green
Orange
Red
Yellow
Unit Size:
Pack of 10
Pack of 150
Description
- Imprinted with hazard symbols and substances of content in German, English and French language
- Writing space
- Abrasion-resistant printing
- Closure color matches color of ink for easy identification of media
- Leak-proof
- Completely emptied by sliding delivery tube, even when inverted.
Specifications
Specifications
| Capacity (Metric) | 250 mL |
| Color | Natural |
| Diameter (Metric) Inner Neck | 30.8 mm |
| Material | LDPE |
| No. per Pack | 10 |
| Quantity | 150 Pk. |
| Closure Color | Red |
| Mouth | Wide |
| Shape | Round |
| Height (Metric) | 120 mm |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction