missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LDOC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA541537
This item is not returnable.
View return policy
Description
Peptide sequence: MVDELVLLLH ALLMRHRALS IENSQLMEQL RLLVCERASL LRQVRPPSCP Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Rabbit: 100%; Rat: 79%.
LDOC1 contains a leucine zipper-like motif and a proline-rich region that shares marked similarity with an SH3-binding domain. The protein localizes to the nucleus and is down-regulated in some cancer cell lines. It is thought to regulate the transcriptional response mediated by the nuclear factor kappa B (NF-kappaB). The gene has been proposed as a tumor suppressor gene whose protein product may have an important role in the development and/or progression of some cancers.The protein encoded by this gene contains a leucine zipper-like motif and a proline-rich region that shares marked similarity with an SH3-binding domain. The protein localizes to the nucleus and is down-regulated in some cancer cell lines. It is thought to regulate the transcriptional response mediated by the nuclear factor kappa B (NF-kappaB). The gene has been proposed as a tumor suppressor gene whose protein product may have an important role in the development and/or progression of some cancers.
Specifications
| LDOC1 | |
| Polyclonal | |
| Unconjugated | |
| Ldoc1 | |
| 7-Mar; BCUR1; breast cancer, up-regulated 1; Gm366; Ldoc1; LDOC1 regulator of NFKB signaling; LDOC1, regulator of NFKB signaling; Leucine zipper; leucine zipper downregulated in cancer; leucine zipper down-regulated in cancer 1; leucine zipper protein down-regulated in cancer cells; leucine zipper, down-regulated in cancer 1; Mar7; Mart7; Protein LDOC1; RGD1565644 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 23641 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O95751 | |
| Ldoc1 | |
| Synthetic peptide directed towards the N-terminal of human LDOC1 (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction