missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LDOC1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99219-50ul
This item is not returnable.
View return policy
Description
LDOC1 Polyclonal antibody specifically detects LDOC1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| LDOC1 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:500-1:2000 | |
| BCUR1, breast cancer, up-regulated 1, Leucine zipper protein down-regulated in cancer cells, leucine zipper, down-regulated in cancer 1, Mar7, Mart7, protein LDOC1 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human LDOC1. | |
| 50 μg | |
| Cell Cycle and Replication | |
| 23641 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction