missing translation for 'onlineSavingsMsg'
Learn More

LDN-212854, MedChemExpress

Product Code. 16476908
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16476908 10 mg 10mg
16472967 50 mg 50mg
16409447 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16476908 Supplier Medchem Express Supplier No. HY15897
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress LDN-212854 is a bone morphogenetic protein (BMP) inhibitor that potently inhibits ALK2 (IC50: 1.3 nM). LDN-212854 also inhibits ALK1 (IC50: 2.40 nM). LDN-212854 can be used in the research of fibrodysplasia ossificans progressive and cancers, such as hepatocellular carcinoma (HCC).

LDN-212854 is a bone morphogenetic protein (BMP) inhibitor that potently inhibits ALK2 (IC50: 1.3 nM). LDN-212854 also inhibits ALK1 (IC50: 2.40 nM). LDN-212854 can be used in the research of fibrodysplasia ossificans progressive and cancers, such as hepatocellular carcinoma (HCC).

Specifications

Chemical Name or Material LDN-212854
CAS 1432597-26-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C25H22N6
Quantity 10 mg
Solubility Information DMSO : ≥ 30 mg/mL (73.80 mM)
SMILES C12=C(C3=C4N=CC(C5=CC=C(N6CCNCC6)C=C5)=CN4N=C3)C=CC=C1N=CC=C2
Molecular Weight (g/mol) 406.48
Formula Weight 406.48
Percent Purity 99.65%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.