missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ LDN 193189 in solution
Description
LDN 193189 in solution is a convenient and ready-to-use 10 mM sterile-filtered solution of LDN 193189 pre-dissolved in water. A potent and selective ALK2 and ALK3 inhibitor (IC50 values are 5 nM and 30 nM, respectively); inhibits BMP4-mediated Smad1/5/8 activation. Promotes neural induction of hPSCs in combination with SB 431542 (Cat. No. 1614). Also induces differentiation of hPSCs into nociceptive sensory neurons in combination with SB 431542 (Cat. No. 1614), SU 5402 (Cat. No. 3300), CHIR 99021 (Cat. No. 4423) and DAPT (Cat. No. 2634).
Specifications
Specifications
| Chemical Name or Material | 4-[6-[4-(1-Piperazinyl)phenyl]pyrazolo[1,5-a]pyrimidin-3-yl]quinoline dihydrochloride |
| CAS | 1435934-00-1 |
| Quantity | 6 mg |
| Target | BMP and Other Activin Receptor Inhibitors |
| Molecular Formula | C25H22N6.2HCl |
| Purity | 0.97 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?