missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ LDN 193189 dihydrochloride
GREENER_CHOICE

Product Code. 18707806
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
Product Code. Quantity unitSize
18707806 10 mg 10mg
18787766 50 mg 50mg
18075802 1 mg 1mg
3 options
This item is not returnable. View return policy

Product Code. 18707806

Brand: Tocris Bioscience™ 6053/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective ALK2 and ALK3 inhibitor; inhibits BMP4 signaling; promotes neural induction of hPSCs

LDN 193189 dihydrochloride is a potent and selective ALK2 and ALK3 inhibitor (IC50 values are 5 and 30 nM, respectively); inhibits BMP4-mediated Smad1/5/8 activation. Exhibits >200-fold selectivity for BMP signaling over TGF-β signaling. Also exhibits selectivity over AMPK, PDGFR and MAPK signaling. Promotes neural induction of hPSCs in combination with SB 431542 (Cat.No. 1614). Also induces differentiation of hPSCs into nociceptive sensory neurons in combination with SB 431542 (Cat.No. 1614), SU 5402 (Cat.No.3300), CHIR 99021 (Cat.No. 4423) and DAPT (Cat.No. 2634).

LDN 193189 synthesized to Ancillary Material Grade also available.

For more information about how LDN 193189 dihydrochloride may be used, see our protocols: Generation of β cells from hPSCs, Generating Midbrain Dopaminergic Neurons from hPSCs, Accelerated Induction of Cortical Neurons from hiPSCs .

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-[6-[4-(1-Piperazinyl)phenyl]pyrazolo[1,5-a]pyrimidin-3-yl]quinoline dihydrochloride
CAS 1435934-00-1
Synonym 4-[6-[4-(1-Piperazinyl)phenyl]pyrazolo[1,5-a]pyrimidin-3-yl]quinoline dihydrochloride
Quantity 10 mg
Solubility Soluble to 50mM in water and to 20mM in DMSO
Target BMP and Other Activin Receptor Inhibitors
Molecular Formula C25H22N60.2 HCl
Formula Weight 479.4
Physical Form Solid
Color Orange
Product Type Bioactive Small Molecule
Purity 98%
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.