missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LDLRAP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112998
This item is not returnable.
View return policy
Description
LDLRAP1 Polyclonal Antibody for Western Blot, IHC (P), ELISA
The protein encoded by this gene is a cytosolic protein which contains a phosphotyrosine binding (PTD) domain. The PTD domain has been found to interact with the cytoplasmic tail of the LDL receptor. Mutations in this gene lead to LDL receptor malfunction and cause the disorder autosomal recessive hypercholesterolaemia.
Specifications
| LDLRAP1 | |
| Polyclonal | |
| Unconjugated | |
| LDLRAP1 | |
| AA691260; Arh; Arh1; ARH2; Autosomal recessive hypercholesterolemia protein; autosomal recessive hypercholesterolemia protein homolog; DKFZp586D0624; FHCB1; FHCB2; LDL receptor adaptor protein; LDLRAP1; Low density lipoprotein receptor adapter protein 1; low density lipoprotein receptor adaptor protein 1; MGC34705; RGD1563417; RP11-70P17.2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100017, 26119 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5SW96, Q8C142 | |
| LDLRAP1 | |
| Recombinant Human Low density lipoprotein receptor adapter protein 1 protein (59-308AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction