missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LDLRAD2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101654
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total LDLRAD2.
The exact functions of this protein remain unknown.
Specifications
| LDLRAD2 | |
| Polyclonal | |
| Unconjugated | |
| LDLRAD2 | |
| LDLRAD2; low density lipoprotein receptor A domain containing 2; low density lipoprotein receptor class A domain containing 2; low-density lipoprotein receptor class A domain-containing protein 2; lrad2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 401944 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q5SZI1 | |
| LDLRAD2 | |
| A synthesized peptide derived from human LDLRAD2(Accession Q5SZI1), corresponding to amino acid residues P129-C179. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction