missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LDLR Antibody (2B10), Novus Biologicals™
Rabbit Monoclonal Antibody
£269.00 - £406.00
Specifications
| Antigen | LDLR |
|---|---|
| Clone | 2B10 |
| Dilution | Flow Cytometry 1:20-1:200, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:20-1:200 |
| Applications | Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30027630
|
Novus Biologicals
NBP3-26172-50ul |
50 μL |
£269.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30028768
|
Novus Biologicals
NBP3-26172-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LDLR Monoclonal antibody specifically detects LDLR in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, ImmunocytochemistrySpecifications
| LDLR | |
| Flow Cytometry 1:20-1:200, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| FH, FHC, LDL receptor, LDLCQ2, low density lipoprotein receptor, low-density lipoprotein receptor, low-density lipoprotein receptor class A domain-containing protein 3 | |
| A synthesized peptide derived from Human LDLR [UniProt P01130] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 2B10 | |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 3949 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title