missing translation for 'onlineSavingsMsg'
Learn More

LDLR Antibody (263123) [Janelia Fluor™ 585], R&D Systems™

Product Code. 30171265 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30171265 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30171265 Supplier Novus Biologicals Supplier No. FAB2255JF585
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rat Monoclonal Antibody

LDLR Monoclonal antibody specifically detects LDLR in Mouse samples. It is validated for Flow Cytometry,Western Blot,CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen LDLR
Applications Flow Cytometry, Western Blot, CyTOF
Classification Monoclonal
Clone 263123
Conjugate Janelia Fluor 585
Formulation 50mM Sodium Borate
Gene Alias FH, FHC, LDL receptor, LDLCQ2, low density lipoprotein receptor, low-density lipoprotein receptor, low-density lipoprotein receptor class A domain-containing protein 3
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse LDL R, Ala22-Arg790 (Ala23Val, Cys27Gly), Accession # Q6GTJ9
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3949
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2a
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.