missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LDLR Antibody (032), DyLight 350, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90523UV
This item is not returnable.
View return policy
Description
LDLR Monoclonal specifically detects LDLR in Mouse samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| LDLR | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| No cross-reactivity in ELISA with: Human LDLR/ LDL Receptor | |
| Store at 4°C in the dark. | |
| IgG |
| Flow Cytometry, ELISA, Immunofluorescence | |
| 32 | |
| Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
| FH, FHC, LDL receptor, LDLCQ2, low density lipoprotein receptor, low-density lipoprotein receptor, low-density lipoprotein receptor class A domain-containing protein 3 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse LDLR (Met 1-Arg 790; NP_034830.2). | |
| 0.1 mL | |
| Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction | |
| 3949 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction