missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LDHD Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584712
This item is not returnable.
View return policy
Description
Immunogen sequence: CAFPSVQAAV DSTVHILQAA VPVARIEFLD EVMMDACNRY SKLNCLVAPT LFLEFHGSQQ ALEEQLQRTE EIVQQNGASD FSWA Highest antigen sequence indentity to the following orthologs: Mouse - 87%, Rat - 89%.
LDHD belongs to the D-isomer specific 2-hydroxyacid dehydrogenase family. The similar protein in yeast has both D-lactate and D-glycerate dehydrogenase activities. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified.
Specifications
| LDHD | |
| Polyclonal | |
| Unconjugated | |
| LDHD | |
| 4733401P21Rik; Ac2-202; D8Bwg1320e; D-lactate dehydrogenase; DLD; lactate dehydrogenase D; LDHD; Probable D-lactate dehydrogenase, mitochondrial | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 197257 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry, Western Blot | |
| 0.6 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86WU2 | |
| LDHD | |
| Recombinant protein corresponding to Human LDHD. Recombinant protein control fragment (Product #RP-105384). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction