missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LDHC Polyclonal Antibody
Goat Polyclonal Antibody
Brand: Invitrogen™ PA518780
This item is not returnable.
View return policy
Description
This antibody is predicted to react with human based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1,000.
Lactate dehydrogenase C catalyzes the conversion of L-lactate and NAD to pyruvate and NADH in the final step of anaerobic glycolysis. LDHC is testis-specific and belongs to the lactate dehydrogenase family. Two transcript variants have been detected which differ in the 5' untranslated region.
Specifications
| LDHC | |
| Polyclonal | |
| Unconjugated | |
| LDHC | |
| Cancer/testis antigen 32; CT32; lactate dehydrogenase 3, C chain, sperm specific; lactate dehydrogenase C; lactate dehydrogenase c variant 1; lactate dehydrogenase c variant 3; lactate dehydrogenase c variant 4; lactate dehydrogenase c variant 5; lactate dehydrogenase C4; lactate dehydrogenase-C; lactate dehydrogenase-X; LDH testis subunit; Ldh3; Ldh-3; LDHC; LDH-C; Ldhc4; LDH-C4; LDHX; Ldh-x; L-lactate dehydrogenase C; L-lactate dehydrogenase C chain; MGC111073; MGC127715 protein; testis secretory sperm-binding protein Li 234P | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 16833, 3948 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P00342, P07864 | |
| LDHC | |
| Synthetic peptide sequence (DSDKEHWKNVHKQ) corresponding to the internal amino acids of LDHC (aa 221-234). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction