missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LDHC Polyclonal Antibody
Description
This antibody is predicted to react with human based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
Lactate dehydrogenase C catalyzes the conversion of L-lactate and NAD to pyruvate and NADH in the final step of anaerobic glycolysis. LDHC is testis-specific and belongs to the lactate dehydrogenase family. Two transcript variants have been detected which differ in the 5' untranslated region.
Specifications
Specifications
| Antigen | LDHC |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | LDHC |
| Gene Accession No. | P00342 |
| Gene Alias | Cancer/testis antigen 32; CT32; lactate dehydrogenase 3, C chain, sperm specific; lactate dehydrogenase C; lactate dehydrogenase c variant 1; lactate dehydrogenase c variant 3; lactate dehydrogenase c variant 4; lactate dehydrogenase c variant 5; lactate dehydrogenase C4; lactate dehydrogenase-C; lactate dehydrogenase-X; LDH testis subunit; Ldh3; Ldh-3; LDHC; LDH-C; Ldhc4; LDH-C4; LDHX; Ldh-x; L-lactate dehydrogenase C; L-lactate dehydrogenase C chain; MGC111073; MGC127715 protein; testis secretory sperm-binding protein Li 234P |
| Gene Symbols | LDHC |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?