missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LDHB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100012
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total LDHB.
This gene encodes an enzyme which catalyzes the reversible conversion of lactate and pyruvate, and NAD and NADH, in the glycolytic pathway. Mutations in this gene are associated with lactate dehydrogenase B deficiency. Pseudogenes have been identified on the X chromosome and on chromosome. Multiple alternatively spliced variants, encoding the same protein, have been identified.
Specifications
| LDHB | |
| Polyclonal | |
| Unconjugated | |
| LDHB | |
| AI790582; epididymis secretory protein Li 281; H(B)-type lactate dehydrogenase; HEL-S-281; H-Ldh; lactate dehydrogenase B; lactate dehydrogenase H chain; lactate dehydrogenase-B; lactate dehydrogenase-H; lactate dehydrogenase-H (, EC 1.1.1.27); Lactate dehydrogenease B; LDH heart subunit; Ldh2; Ldh-2; LDHB; LDH-B; LDHBD; LDHH; LDH-H; L-lactate dehydrogenase B chain; Renal carcinoma antigen NY-REN-46; similar to lactate dehydrogenase B; testicular secretory protein Li 25; TRG-5 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 16832, 24534, 3945 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P07195, P16125, P42123 | |
| LDHB | |
| A synthesized peptide derived from human LDHB(Accession P07195), corresponding to amino acid residues M1-L45. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction