missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LDHB Recombinant Rabbit Monoclonal Antibody (6B5I2)

LDHB Recombinant Rabbit Monoclonal Antibody (ARC1217)
Brand: Invitrogen™ MA535655
This item is not returnable.
View return policy
Description
LDHB Recombinant Monoclonal Antibody for Western Blot, ICC/IF
This gene encodes an enzyme which catalyzes the reversible conversion of lactate and pyruvate, and NAD and NADH, in the glycolytic pathway. Mutations in this gene are associated with lactate dehydrogenase B deficiency. Pseudogenes have been identified on the X chromosome and on chromosome. Multiple alternatively spliced variants, encoding the same protein, have been identified.
Specifications
| LDHB | |
| Recombinant Monoclonal | |
| 0.6 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P07195, P42123 | |
| LDHB | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human LDHB (P07195). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 6B5I2 | |
| Unconjugated | |
| LDHB | |
| AI790582; epididymis secretory protein Li 281; H(B)-type lactate dehydrogenase; HEL-S-281; H-Ldh; lactate dehydrogenase B; lactate dehydrogenase H chain; lactate dehydrogenase-B; lactate dehydrogenase-H; lactate dehydrogenase-H (, EC 1.1.1.27); Lactate dehydrogenease B; LDH heart subunit; Ldh2; Ldh-2; LDHB; LDH-B; LDHBD; LDHH; LDH-H; L-lactate dehydrogenase B chain; Renal carcinoma antigen NY-REN-46; similar to lactate dehydrogenase B; testicular secretory protein Li 25; TRG-5 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 24534, 3945 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction