missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LDHAL6B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562885
This item is not returnable.
View return policy
Description
Immunogen sequence: TVPVVRASQR VSSVGANFLC LGMALCPRQA TRIPLNGTWL FTPVSKMATV KSELIERFTS EKPV Highest antigen sequence identity to the following orthologs: Mouse - 48%, Rat - 30%.
LDHAL6B belongs to the LDH/MDH superfamily. LDH family. The specific function of the protein remains unknown.
Specifications
| LDHAL6B | |
| Polyclonal | |
| Unconjugated | |
| LDHAL6B | |
| 4933402O15Rik; AI326310; lactate dehydrogenase A like 6B; lactate dehydrogenase A-like 6; lactate dehydrogenase A-like 6B; LDH6B; Ldhal; LDHAL6; Ldhal6b; LDHL; L-lactate dehydrogenase A-like 6B; testicular tissue protein Li 105 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 92483 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BYZ2 | |
| LDHAL6B | |
| Recombinant protein corresponding to Human LDHAL6B. Recombinant protein control fragment (Product #RP-101462). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction