missing translation for 'onlineSavingsMsg'
Learn More

LDC000067, MedChemExpress

Product Code. 16496888
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16496888 10 mg 10mg
16402957 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16496888 Supplier Medchem Express Supplier No. HY15878
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress LDC000067 is a highly specific CDK9 inhibitor with an IC50 value of 44±10 nM in vitro.

LDC000067 is a highly specific CDK9 inhibitor with an IC50 value of 44±10 nM in vitro.

Specifications

Chemical Name or Material LDC000067
CAS 1073485-20-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C18H18N4O3S
Quantity 10 mg
Synonym LDC067
Solubility Information DMSO : ≥ 47 mg/mL (126.88 mM)
SMILES O=S(CC1=CC=CC(NC2=NC=NC(C3=CC=CC=C3OC)=C2)=C1)(N)=O
Molecular Weight (g/mol) 370.43
Formula Weight 370.43
Percent Purity 98.58%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.