missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ LDC 000067
Description
LDC 000067 is a potent and selective CDK9 inhibitor (IC50 = 44 nM at CDK9-cyclin T1). Inhibits other CDKs in the micromolar range (IC50 values are 2.4, 5.5, 9.2, >10 and >10 μM for CDK2-cyclin A, CDK1-cyclin B1, CDK4-cyclin D1, CDK6-cyclin D3 and CDK7-cyclin H-MAT1, respectively). Inhibits P-TEFb-dependent in vitro transcription. Induces apoptosis in vitro and in vivo in combination with BI 894999.
Specifications
Specifications
| Chemical Name or Material | 3-[[6-(2-Methoxyphenyl)-4-pyrimidinyl]amino]benzenemethanesulfonamide |
| CAS | 1073485-20-7 |
| Quantity | 5 mg |
| Molecular Formula | C18H18N4O3S |
| Formula Weight | 370.43 |
| Purity | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?