missing translation for 'onlineSavingsMsg'
Learn More

LDB2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-19341

Product Code. 18160222

  • £494.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

LDB2 Polyclonal specifically detects LDB2 in Human, Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LDB2
Polyclonal
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Carboxyl-terminal LIM domain-binding protein 1, CLIM-1, CLIM1LDB1, LDB-2, LIM binding domain 2, LIM domain binding 2, LIM domain-binding factor CLIM1, LIM domain-binding factor-2, LIM domain-binding protein 2
Rabbit
43 kDa
0.1 mL
Stem Cell Markers
9079
Human, Mouse
IgG
Western Blot
0.73mg/mL
Western Blot 1:500-1:3000
O43679
LDB2
Recombinant protein encompassing a sequence within the center region of human LDB2. The exact sequence is proprietary.
Affinity Purified
RUO
Primary
Human, Mouse. Species reactivity based on sequence identity: Rhesus Monkey (100%).
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only